Thermo Scientific Chemicals

Manganese(II) carbonate, Mn 44% min, Thermo Scientific Chemicals

Catalog number: 014324.36
500 g, Each
Thermo Scientific Chemicals

Manganese(II) carbonate, Mn 44% min, Thermo Scientific Chemicals

Catalog number: 014324.36
500 g, Each
Quantity
Catalog number: 014324.36
also known as 014324-36
Price (USD)
Price: 54.00
Online price: 45.65
Your price:
Quantity
-

Chemical Identifiers

CAS
598-62-9
IUPAC Name
manganese(2+) carbonate
Molecular Formula
CMnO3
InChI Key
XMWCXZJXESXBBY-UHFFFAOYSA-L
SMILES
[Mn++].[O-]C([O-])=O
Assay (unspecified)
44% Mn min

Description

Pigment, varnish drierManganese(II) carbonate is used as an additive to plant fertilizers, glaze colorant in ceramics, pigment and drier for varnishes. It is also used as an ingredient in welding rod, animal feed additive as well as to prepare other manganese salts and pharmaceuticals. It is also used in medicine as a hematinic and in health foods, in flux and in concrete stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text