Thermo Scientific Chemicals

Tin(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 014476.14
25 g, Each
Thermo Scientific Chemicals

Tin(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 014476.14
25 g, Each
Quantity
Catalog number: 014476.14
Price (USD)
Price: 556.00
Online price: 472.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10294-70-9
IUPAC Name
λ²-tin(2+) diiodide
Molecular Formula
I2Sn
InChI Key
JTDNNCYXCFHBGG-UHFFFAOYSA-L
SMILES
[Sn++].[I-].[I-]
Appearance (Color)
Red
Residual water
< 100 ppm
Total Metal Impurities
0.001% max.
Particle Size
-10 Mesh
Form
Beads

Description

Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3 which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text