Holmium(III)-nitratpentahydrat, REacton™, 99.9 % (REO), Thermo Scientific Chemicals
Holmium(III)-nitratpentahydrat, REacton™, 99.9 % (REO), Thermo Scientific Chemicals
Holmium(III)-nitratpentahydrat, REacton™, 99.9 % (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium(III)-nitratpentahydrat, REacton™, 99.9 % (REO), Thermo Scientific Chemicals

CAS: 14483-18-2 | H10HoN3O14 | 441.017 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
50 g
Katalognummer 014588.18
Preis (EUR)
405,00
Each
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
405,00
Each
Chemikalien-Kennzeichnungen
CAS14483-18-2
IUPAC Nameholmium(3+) pentahydrate trinitrate
Molecular FormulaH10HoN3O14
InChI KeyBJUGWWDCFYEYOA-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Ho+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Rare Earth Oxide Impurities0.1% max.
Holmium(III) nitrate pentahydrate is used to prepare holmium-beta-cyclodextrin complex by reacting with beta- cyclodextrin, which posses potential pharmacological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only