Thermo Scientific Chemicals

Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014603.14
25 g, Each
Thermo Scientific Chemicals

Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014603.14
25 g, Each
Quantity
Catalog number: 014603.14
also known as 014603-14
Price (USD)
Price: 58.90
Online price: 50.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13517-12-9
IUPAC Name
samarium(3+) hexahydrate tribromide
Molecular Formula
Br3H12O6Sm
InChI Key
LXJDCZVUWJNXRL-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text