Thermo Scientific Chemicals

Barium chromate, 98%, Thermo Scientific Chemicals

Catalog number: 014669.A1
1 kg, Each
Thermo Scientific Chemicals

Barium chromate, 98%, Thermo Scientific Chemicals

Catalog number: 014669.A1
1 kg, Each
Quantity
Catalog number: 014669.A1
also known as 014669-A1
Price (USD)
Quantity
-

Chemical Identifiers

CAS
10294-40-3
IUPAC Name
barium(2+) dioxochromiumbis(olate)
Molecular Formula
BaCrO4
InChI Key
QFFVPLLCYGOFPU-UHFFFAOYSA-N
SMILES
[Ba++].[O-][Cr]([O-])(=O)=O
Form
Powder or crystalline powder or crystals
Appearance (Color)
Pale yellow to yellow
Assay from Suppliers CofA
≥97.5%

Description

Barium chromate is used as a pigment, oxidizing agent, colorant in glass, and sulfate scavenger in chromium electroplating baths, ceramics, and porcelain. It acts as a corrosion inhibitor in jointing pastes and metal primers. Further, it is used in fuses, safety matches, ignition control devices and high-temperature batteries. In addition to this, it is used as a carrier for the chromium ions and useful as a burn rate modifier in pyrotechnic compositions. It is involved as a catalyst in the alkane dehydration reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text