Natriumhexametaphosphat, technischer Gütegrad, Thermo Scientific Chemicals
Natriumhexametaphosphat, technischer Gütegrad, Thermo Scientific Chemicals
Natriumhexametaphosphat, technischer Gütegrad, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Natriumhexametaphosphat, technischer Gütegrad, Thermo Scientific Chemicals

CAS: 10124-56-8 | Na6O18P6
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 kg
5 kg
Katalognummer 014706.A1
auch als 014706-A1 bezeichnet
Preis (EUR)
87,65
Exklusiv online
97,40
Ersparnis 9,75 (10%)
Each
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
87,65
Exklusiv online
97,40
Ersparnis 9,75 (10%)
Each
Chemikalien-Kennzeichnungen
CAS10124-56-8
IUPAC Namehexasodium hexaoxocyclohexaphosphoxane-2,4,6,8,10,12-hexakis(olate)
Molecular FormulaNa6O18P6
InChI KeyGCLGEJMYGQKIIW-UHFFFAOYSA-H
SMILES[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P1(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)O1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
FormPowder
Assay67.5-69.7% (Phosphorus pentoxide-P2O5)
Assay (unspecified)67.5-69.7% (Phosphorus pentoxide-P2O5)
pH5.8-7.3 (1% aq. solution)
Sodium hexametaphosphate is a food additive used in dairy product, canned milk, packaged egg whites, ice cream, seafood and meat processing. It is also used as a sequestrant and water softening agent and detergents. It is an active ingredient in toothpastes as an anti-staining agent. It prevents the steel corrosion. Further, it is used in building, water treatment and in photographic developing. In addition, it is used as a retarder in dental alginate impression materials and as a thinner for petroleum drilling fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only