Thermo Scientific Chemicals
Graphite powder, natural, high purity, -200 mesh, 99.9999% (metals basis), Thermo Scientific Chemicals
Catalog number: 014734.TA
28 g, Each
Thermo Scientific Chemicals
Graphite powder, natural, high purity, -200 mesh, 99.9999% (metals basis), Thermo Scientific Chemicals
Catalog number: 014734.TA
28 g, Each
Quantity
Appearance (Color)
Black
Form
Powder
Total Metal Impurities
≤0.0002%
Description
Graphite powder is used to make crucibles, lead pencils, electrodes, lubricants, stove polish, brake linings, batteries, glass manufacture, conductive coatings, and for foundry facings. It is used in the synthesis of single sheet graphene nanoflakes and graphite based magnetorheological elastomers.