Thermo Scientific Chemicals

Barium silicon oxide, Thermo Scientific Chemicals

Catalog number: 015122.36
500 g, Each
Thermo Scientific Chemicals

Barium silicon oxide, Thermo Scientific Chemicals

Catalog number: 015122.36
500 g, Each
Quantity
Catalog number: 015122.36
also known as 015122-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
13255-26-0
IUPAC Name
barium(2+) oxosilanebis(olate)
Molecular Formula
BaO3Si
InChI Key
HMOQPOVBDRFNIU-UHFFFAOYSA-N
SMILES
[Ba++].[O-][Si]([O-])=O
Assay (unspecified)
34% BaO min

Description

Barium silicate films that are suitable for use as transmission media in integrated optical circuits. It is shown that the films, which can be produced with a wide range of refractive indices by suitable selection of the ratio of the target constituents, exhibit low optical attenuation. Hence it is used for integrated optical circuits. It is used in ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text