Europium(III)-nitrathexahydrat, 99.9 % (REO), Thermo Scientific Chemicals, REacton™
Europium(III)-nitrathexahydrat, 99.9 % (REO), Thermo Scientific Chemicals, REacton™
Europium(III)-nitrathexahydrat, 99.9 % (REO), Thermo Scientific Chemicals, REacton™
Thermo Scientific Chemicals

Europium(III)-nitrathexahydrat, 99.9 % (REO), Thermo Scientific Chemicals, REacton™

CAS: 10031-53-5 | EuH12N3O15 | 446.066 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
2 g
10 g
50 g
Katalognummer 015290.09
auch als 015290-09 bezeichnet
Preis (EUR)
333,00
Each
Menge:
10 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
333,00
Each
Chemikalien-Kennzeichnungen
CAS10031-53-5
IUPAC Nameeuropium(3+) hexahydrate trinitrate
Molecular FormulaEuH12N3O15
InChI KeyJVYYYCWKSSSCEI-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Rare Earth Oxide Impurities0.1% max.
Europium(III) nitrate hexahydrate is used in the preparation of pharmaceuticals, agrochemicals and other chemical products. It is used as a catalyst and a component of specialized steels. It is also used in the synthesis of light emitting nanomaterials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only