Lanthan(III)-acetat-Sesquihydrat, 99.9 % (REO), Thermo Scientific Chemicals
Lanthan(III)-acetat-Sesquihydrat, 99.9 % (REO), Thermo Scientific Chemicals
Lanthan(III)-acetat-Sesquihydrat, 99.9 % (REO), Thermo Scientific Chemicals
Lanthan(III)-acetat-Sesquihydrat, 99.9 % (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthan(III)-acetat-Sesquihydrat, 99.9 % (REO), Thermo Scientific Chemicals

CAS: 25721-92-0 | C12H24La2O15 | 686.12 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
Katalognummer 015299.36
auch als 015299-36 bezeichnet
Preis (EUR)
247,00
Each
Menge:
500 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
247,00
Each
Chemikalien-Kennzeichnungen
CAS25721-92-0
IUPAC Namedilanthanum(3+) hexaacetate trihydrate
Molecular FormulaC12H24La2O15
InChI KeyRBPQRWPSWRTKEC-UHFFFAOYSA-H
SMILESO.O.O.[La+3].[La+3].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
FormLump/Powder
Total Rare Earth Oxide Impurities>99.9% REO
Lanthanum(III) acetate sesquihydrate is used in specialty glass and in water treatment. It is used in the preparation of phenolate-bridged dilanthanum(III) complex, which is considered as a model for metalloproteins in applied chemistry. Further, it is used to prepare nanoparticulate porous lanthanum oxyfluoride (LaOF) films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only