Diethylether, wasserfrei, ACS, 99 % min., mit BHT stab., Thermo Scientific Chemicals
Diethylether, wasserfrei, ACS, 99 % min., mit BHT stab., Thermo Scientific Chemicals
Diethylether, wasserfrei, ACS, 99 % min., mit BHT stab., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylether, wasserfrei, ACS, 99 % min., mit BHT stab., Thermo Scientific Chemicals

CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
500 g
6 x 500 g
Katalognummer 016767.62
auch als 016767-62 bezeichnet
Preis (EUR)
277,00
Each
Menge:
6 x 500 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
277,00
Each
Chemikalien-Kennzeichnungen
CAS60-29-7
IUPAC Nameethoxyethane
Molecular FormulaC4H10O
InChI KeyRTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILESCCOCC
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormClear liquid
Color (APHA)≤10
Assay (GC)≥99%
CommentStabilized with BHT
Free acid (titration)≤0.0002meq/g
Mehr anzeigen
Solvent for U.V. spectrophotometric and it is also important as solvent in production of cellulose plastics such as cellulose acetate. It is also common solvent for Grignard reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only