Thermo Scientific Chemicals

Scandium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 018677.06
5 g, Each
Thermo Scientific Chemicals

Scandium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 018677.06
5 g, Each
Quantity
Catalog number: 018677.06
also known as 018677-06
Price (USD)
Price: 869.00
Online price: 738.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10361-84-9
IUPAC Name
scandium(3+) trichloride
Molecular Formula
Cl3Sc
InChI Key
DVMZCYSFPFUKKE-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Sc+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Scandium(III) chloride is a Prins cyclization catalyst. Used to form an intermetallic nickel-scandium compound film by potentiostatic cathodic reduction from molten salt mixtures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text