Thermo Scientific Chemicals

n-Butyl acetate, Semiconductor Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 019395.K7
4 L, Each
Thermo Scientific Chemicals

n-Butyl acetate, Semiconductor Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 019395.K7
4 L, Each
Quantity
Catalog number: 019395.K7
also known as 019395-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-86-4
IUPAC Name
butyl acetate
Molecular Formula
C6H12O2
InChI Key
DKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILES
CCCCOC(C)=O
Form
Liquid
Elemental Analysis
≤10 ppb for all of the following: Al,Ba,Be,Bi,B,Cd,Ca,Cr,Co,Cu,Ga,Ge,Au,Fe,Pb,Li,Mg,Mn,Mo,Ni,Nb,K,Si,Ag
Water Content (Karl Fischer Titration)
≤500 ppm
Particle Size
Particles 1.0 micron: ≤5 particles/ml
Assay (GC)
≥99.0%

Description

n-Butyl acetate is widely used as a solvent for cellulose nitrate, resins, polymers, oils and fats. It is used in paints, coatings, printing inks, aerosol sprays and cosmetics. It is used as a synthetic fruit flavoring agent in foods like candy, ice cream, cheeses and baked food items. In the pharmaceutical industry, it is used for extraction and as processing solvent. It is an ingredient of cleaners, essences and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text