Kupfer(II)-acetat, wasserfrei, Thermo Scientific Chemicals
Kupfer(II)-acetat, wasserfrei, Thermo Scientific Chemicals
Kupfer(II)-acetat, wasserfrei, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kupfer(II)-acetat, wasserfrei, Thermo Scientific Chemicals

CAS: 142-71-2 | C4H6CuO4 | 181.634 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
500 g
Katalognummer 019417.14
auch als 019417-14 bezeichnet
Preis (EUR)
110,00
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
110,00
Each
Chemikalien-Kennzeichnungen
CAS142-71-2
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)97% min
Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, textile dyeing, skin conditioner in cosmetics, antioxidant and stabilizer in food grade polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only