Lactic acid, 88.0-92.0%, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Lactic acid, 88.0-92.0%, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, and E.P specifications.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 mL
4 L
19 L
Katalognummer 0200-07
Preis (EUR)
-
Menge:
19 L
This product is GMP Manufactured and meets J.P., U.S.P, and E.P specifications.
This product is GMP Manufactured and meets J.P., U.S.P, and E.P specifications.
Specifications
Siedepunkt122°C
CAS50-21-5
Chemischer Name oder MaterialLactic acid
FarbeFarblos
SummenformelH3CCH(OH)COOH
VerpackungPoly Pail (Jerrican)
Synonym2-Hydroxypropionic acid, 2-Hydroxypropanoic acid, DL-Lactic acid
GüteMulti-Kompendium
Molekulargewicht (g/mol)90.08
Reinheit (%)88 to 92%
Menge19 L
Unit SizeEach