Thermo Scientific Chemicals

Methyl malonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 020823.14
25 g, Each
Thermo Scientific Chemicals

Methyl malonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 020823.14
25 g, Each
Quantity
Catalog number: 020823.14
also known as 020823-14
Price (USD)
Price: 208.00
Online price: 179.65
Your price:
Quantity
-

Chemical Identifiers

CAS
37517-81-0
IUPAC Name
methyl 3-chloro-3-oxopropanoate
Molecular Formula
C4H5ClO3
InChI Key
UTBCRHAMJFMIIR-UHFFFAOYSA-N
SMILES
COC(=O)CC(Cl)=O
Assay (GC)
≥96.0%
Refractive Index
1.4310-1.4350 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

Methyl malonyl chloride is used as pharmaceutical intermediate. Used in anti-cancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text