Natriumtrifluormethansulfonat, Thermo Scientific Chemicals
Natriumtrifluormethansulfonat, Thermo Scientific Chemicals
Natriumtrifluormethansulfonat, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Natriumtrifluormethansulfonat, Thermo Scientific Chemicals

CAS: 2926-30-9 | CF3NaO3S | 172.053 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
2 g
10 g
50 g
Katalognummer 020837.18
auch als 020837-18 bezeichnet
Preis (EUR)
288,00
Each
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
288,00
Each
Chemikalien-Kennzeichnungen
CAS2926-30-9
IUPAC Namesodium trifluoromethanesulfonate
Molecular FormulaCF3NaO3S
InChI KeyXGPOMXSYOKFBHS-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C(F)(F)F
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Purity≥95.0%
Assay (unspecified)Sodium (Na): ≥12.7%
FormPowder
Catalyzes the aza Diels-Alder reaction of Danishefsky's diene with imines in water under neutral conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only