Thermo Scientific Chemicals

Ammonium tetrathiotungstate, Thermo Scientific Chemicals

Catalog number: 022373.09
10 g, Each
Thermo Scientific Chemicals

Ammonium tetrathiotungstate, Thermo Scientific Chemicals

Catalog number: 022373.09
10 g, Each
Quantity
Catalog number: 022373.09
also known as 022373-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13862-78-7
IUPAC Name
disulfanylidenetungsten diamine disulfanide
Molecular Formula
H8N2S4W
InChI Key
CCFCMIWYHMKPAO-UHFFFAOYSA-L
SMILES
N.N.[SH-].[SH-].S=[W]=S
Impurity content
0.1% max

Description

Ammonium tetrathiotungstate is used to classic building block for transition metal sulfide cluster compounds was recently used in a low-temperature, solid state synthesis of a new series of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text