Thermo Scientific Chemicals

N,N-Dimethylformamide, HPLC Grade, 99.7+%, Thermo Scientific Chemicals

Catalog number: 022915.K2
1 L, Each
Thermo Scientific Chemicals

N,N-Dimethylformamide, HPLC Grade, 99.7+%, Thermo Scientific Chemicals

Catalog number: 022915.K2
1 L, Each
Quantity
Catalog number: 022915.K2
also known as 022915-K2
Price (USD)
Price: 90.70
Online price: 77.65
Your price:
Quantity
-

Chemical Identifiers

CAS
68-12-2
IUPAC Name
N,N-dimethylformamide
Molecular Formula
C3H7NO
InChI Key
ZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILES
CN(C)C=O
Residual water
0.05%
Residue on Evaporation
5ppm

Description

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is used as a mobile phase in High Performance Liquid Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text