Calciumtetrafluorborathydrat, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calciumtetrafluorborathydrat, Thermo Scientific Chemicals

CAS: 206996-55-6 | B2CaF8 | 213.685 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
Katalognummer 030131.22
auch als 030131-22 bezeichnet
Preis (EUR)
548,00
Each
Menge:
100 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
548,00
Each
Chemikalien-Kennzeichnungen
CAS206996-55-6
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)Ca 12% min
Calcium tetrafluoroborate hydrate is used as pharmaceutical and chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only