Search
Suchen
View additional product information for Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 2;ID (mm), 1 - FAQs (031487.DW, 031487.DR)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.