Search
Suchen
View additional product information for Al-23 Tube (both ends open);OD (mm), 4;ID (mm), 2 - FAQs (031554.EM, 031554.DW)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.