Mineralöl, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mineralöl, Thermo Scientific Chemicals

CAS: 8020-83-5
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 kg
5 x 1 kg
Katalognummer 031911.D9
auch als 031911-D9 bezeichnet
Preis (EUR)
298,65
Exklusiv online
332,00
Ersparnis 33,35 (10%)
Each
Menge:
5 x 1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
298,65
Exklusiv online
332,00
Ersparnis 33,35 (10%)
Each
Chemikalien-Kennzeichnungen
CAS8042-47-5
SpecificationsSpecification SheetDatenblatt
Specific Gravity0.8490 to 0.8650 ( at 25?C)
Viscosity23.90 - 27.01cSt at 40?C
Mineral oil is used as an overlay covering microdrops of culture medium in petri dishes. It is an active ingredient in baby lotions, cold creams, ointments and cosmetics. It is also used in electrical and mechanical industries as thermal fluid in electrical components and non -conductive coolant. It finds application as a transformer oil, as an anti rust agent for blades and as an insulator in high voltage switchgear. Further, it is used to store reactive metals like lithium, sodium, etc and in X-ray crystallography to suspend crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only