Zink-Protoporphyrin, 96 %, Thermo Scientific Chemicals
Zink-Protoporphyrin, 96 %, Thermo Scientific Chemicals
Zink-Protoporphyrin, 96 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zink-Protoporphyrin, 96 %, Thermo Scientific Chemicals

CAS: 15442-64-5 | C34H32N4O4Zn | 626.034 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 mg
50 mg
0.25 g
Katalognummer 032068.01
auch als 032068-01 bezeichnet
Preis (EUR)
715,00
Each
Menge:
0.25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
715,00
Each
Specifications
CAS15442-64-5
Chemischer Name oder MaterialZinc protoporphyrin
FarbeViolett bis Rot
EINECS-Nummer239-455-7
Formelmasse626.04
Mehr anzeigen
Zinc protoporphyrin-9 is a potent, selective heme oxygenase inhibitor, interleukin-1 antagonist, GCS inhibitor, and NOS1, NOS2, and NOS3 inhibitor. In olfactory neuronal cultures, this product has been used to block CO biosynthesis, which lowered cGMP concentrations. Zinc protoporphyrin-9 is an extremely useful tool to elucidate the role of CO in signal transduction and as a neurotransmitter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only