Search
View additional product information for Al-23 Multibore Round Tube, O.D.: 1.2mm, Two 0.3mm Bores, Thermo Scientific Chemicals - FAQs (032565.DT, 032565.QAT)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.