Sulfat, Standardlösung für die Ionenchromatographie, Specpure™, SO4-2 1000 μg/ml, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfat, Standardlösung für die Ionenchromatographie, Specpure™, SO4-2 1000 μg/ml, Thermo Scientific Chemicals

Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 mL
500 mL
Katalognummer 035547.AE
Preis (EUR)
133,65
Exklusiv online
148,00
Ersparnis 14,35 (10%)
Each
Menge:
100 mL
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
133,65
Exklusiv online
148,00
Ersparnis 14,35 (10%)
Each
Specifications
Chemischer Name oder MaterialSulfate Ion chromatography standard solution
MDL-NummerMFCD00145343
NamenshinweisSO4-2 1000μg/mL
Physikalische FormFlüssig
Menge100 mL
Mehr anzeigen
Sulfate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is also used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only