Zinkchlorid, ultratrocken, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Zinkchlorid, ultratrocken, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Zinkchlorid, ultratrocken, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinkchlorid, ultratrocken, 99.999 % (Metallbasis), Thermo Scientific Chemicals

CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
100 g
Katalognummer 035726.14
auch als 035726-14 bezeichnet
Preis (EUR)
303,00
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
303,00
Each
Chemikalien-Kennzeichnungen
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Residual water< 100 ppm
FormPowder
Total Metal Impurities0.001% max.
Appearance (Color)White
Zinc(II) chloride is used as a catalyst, as a dehydrating and condensing agent, soldering flux and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in an organic synthesis, for example in the Friedel-crafts acylation, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only