Thermo Scientific Chemicals

Terbium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035798.03
1 g, Each
Thermo Scientific Chemicals

Terbium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035798.03
1 g, Each
Quantity
Catalog number: 035798.03
also known as 035798-03
Price (USD)
Price: 103.00
Online price: 89.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10042-88-3
IUPAC Name
terbium(3+) hexahydrate trichloride
Molecular Formula
Cl3H12O6Tb
InChI Key
ULJUVCOAZNLCJZ-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]
Appearance (Color)
White
Form
Powder
Residual water
< 100 ppm
Total Rare Earth Oxide Impurities
0.01% max.

Description

Terbium(III) chloride is used generally where Tb3+ ions can be used to check for the presence of microbes. Terbium chloride is applied to the test area, which is then illuminated with UV light. Within minutes, any live endospores present will glow green.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text