Ammoniumtetrafluoroborat, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Ammoniumtetrafluoroborat, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Ammoniumtetrafluoroborat, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammoniumtetrafluoroborat, 99.5 % (Metallbasis), Thermo Scientific Chemicals

CAS: 13826-83-0 | BF4H4N | 104.84 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 g
1 kg
Katalognummer 035831.30
auch als 035831-30 bezeichnet
Preis (EUR)
103,00
Each
Menge:
250 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
103,00
Each
Chemikalien-Kennzeichnungen
CAS13826-83-0
IUPAC Nameammonium tetrafluoroboranuide
Molecular FormulaBF4H4N
InChI KeyPDTKOBRZPAIMRD-UHFFFAOYSA-O
SMILES[NH4+].F[B-](F)(F)F
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
Form-40 Mesh
Total Metal Impurities≤0.5%
Ammonium tetrafluoroborate is used in the metal industry as a high-temperature flux. It is also used as a flame retardant and lubricant in aluminum cutting-oils. Further, it acts as an analytical reagent. In addition, it is used as a catalyst for resin finishing and also finds use in textile printing and dyeing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only