Lithiumsulfat-Monohydrat, ACS, 99.0 % min., Thermo Scientific Chemicals
Lithiumsulfat-Monohydrat, ACS, 99.0 % min., Thermo Scientific Chemicals
Lithiumsulfat-Monohydrat, ACS, 99.0 % min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithiumsulfat-Monohydrat, ACS, 99.0 % min., Thermo Scientific Chemicals

Maximaler Gehalt an Verunreinigungen: Trockenverlust bei 150 °C 13.0 bis 15.0 %, unlösliche Stoffe 0.01 %, Cl 0.002 %, NO3 0.001 %, Schwermetalle (als Pb) 0.001 %, Fe 0.001 %, K 0.05 %, Na 0.05 %
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
2 kg
Katalognummer 036216.36
auch als 036216-36 bezeichnet
Preis (EUR)
196,00
Each
Menge:
500 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
196,00
Each
Chemikalien-Kennzeichnungen
CAS10102-25-7
IUPAC Namedilithium(1+) hydrate sulfate
Molecular FormulaH2Li2O5S
InChI KeyRKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES[Li+].[Li+].O.[O-]S([O-])(=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)99.0% Li₂SO₄ min
Impurity contentCl 0.002%
Impurity contentFe 0.001%
Impurity contentK 0.05%
Impurity contentNa 0.05%
Mehr anzeigen
Lithium sulfate monohydrate is used in solar panels and potential for a new class of battery. It is also used to treat bipolar disorder and in ultrasound-type non-destructive testing. It is also used as an additive for portland cement to speed up the curing. It is also involved as a catalyst in the elimination reaction for the conversion of n-butyl bromide to 1-butene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only