Samarium(III)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Samarium(III)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Samarium(III)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Samarium(III)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals

CAS: 12060-58-1 | Sm2O3
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
50 g
Katalognummer 036278.18
auch als 036278-18 bezeichnet
Preis (EUR)
388,00
Each
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
388,00
Each
Chemikalien-Kennzeichnungen
CAS12060-58-1
IUPAC Namedisamarium(3+) trioxidandiide
Molecular FormulaO3Sm2
InChI KeyFKTOIHSPIPYAPE-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sm+3].[Sm+3]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.1% max.
The biocompatibility of natural samarium (III) oxide, which has previously been used for treatment in bone-related diseases was determined as a first step in its evaluation as a bone implant material. The rates and products of the purely heterogeneous oxidations of C2H6 (g) and C2H4 (g) on Sm2O3 in the presence of O2 (g) were investigated in a very low-pressure flow reactor by on-line molecular beam mass spectrometry, about 1000±100 K. Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. Also, it is used as a neutron absorber in control rods for nuclear power reactors. The oxide catalyzes dehydration of acyclic primary alcohols to aldehydes and ketones. Another use involves preparation of other samarium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only