Thermo Scientific Chemicals

Mercury(II) sulfate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 036286.36
500 g, Each
Thermo Scientific Chemicals

Mercury(II) sulfate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 036286.36
500 g, Each
Quantity
Catalog number: 036286.36
also known as 036286-36
Price (USD)

Chemical Identifiers

CAS
7783-35-9
IUPAC Name
mercury(2+) sulfate
Molecular Formula
HgO4S
InChI Key
DOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES
[Hg++].[O-]S([O-])(=O)=O
Assay (unspecified)
98.0% HgSO₄ min
Comment
Residue after reduction 0.02%
Impurity content
Cl 0.003%
Impurity content
Fe 0.005%
Impurity content
Mercurous mercury (as Hg) 0.15%

Description

Mercury(II) sulfate is used for gold and silver extraction, and to make other mercury compounds. Used as catalyst in the conversion of acetylene to acetaldehyde and used as a electrolyte in primary battery. It is commonly used as a catalyst in oxymercuration-demercuration, a type of electrophilic addition reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text