Antimon(V)-oxid, weißer Klumpen, 99.99 % (Metallbasis), Thermo Scientific Chemicals
Antimon(V)-oxid, weißer Klumpen, 99.99 % (Metallbasis), Thermo Scientific Chemicals
Antimon(V)-oxid, weißer Klumpen, 99.99 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimon(V)-oxid, weißer Klumpen, 99.99 % (Metallbasis), Thermo Scientific Chemicals

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
Katalognummer 036314.30
auch als 036314-30 bezeichnet
Preis (EUR)
785,00
Each
Menge:
250 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
785,00
Each
Chemikalien-Kennzeichnungen
CAS1309-64-4
IUPAC Namediantimony(3+) trioxidandiide
Molecular FormulaO3Sb2
InChI KeyGHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sb+3].[Sb+3]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.1% max.
Flame retardant for wide range of plastics, rubbers, paper and textiles . Catalyst in PET production. Activator in glass industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only