Thermo Scientific Chemicals

Sodium hexafluoroaluminate, 99.4% (metals basis), Thermo Scientific Chemicals

Catalog number: 036345.30
250 g, Each
Thermo Scientific Chemicals

Sodium hexafluoroaluminate, 99.4% (metals basis), Thermo Scientific Chemicals

Catalog number: 036345.30
250 g, Each
Quantity
Catalog number: 036345.30
also known as 036345-30
Price (USD)
Price: 407.00
Online price: 345.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13775-53-6
IUPAC Name
aluminium(3+) trisodium hexafluoride
Molecular Formula
AlF6Na3
InChI Key
REHXRBDMVPYGJX-UHFFFAOYSA-H
SMILES
[F-].[F-].[F-].[F-].[F-].[F-].[Na+].[Na+].[Na+].[Al+3]
Appearance (Color)
White
Form
Lump, ca. 3-12mm
Total Metal Impurities
≤0.6%

Description

Mainly used as fluxing agent in the production of aluminum electrolysis, also used as additive to the wear-resisting of abrasive products, iron alloy and the flux of rimming steel, non-ferrous metal flux, casting deoxidizer, enamel of emulsifier, glass opal agent, welding flux and ceramics filler. Aluminum- metallurgy. This chemical is used as adhesives, cosolvent in enamel industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text