Thermo Scientific Chemicals

D-(-)-Isoascorbic acid, 98%, Thermo Scientific Chemicals

Catalog number: 036366.36
500 g, Each
Thermo Scientific Chemicals

D-(-)-Isoascorbic acid, 98%, Thermo Scientific Chemicals

Catalog number: 036366.36
500 g, Each
Quantity
Catalog number: 036366.36
also known as 036366-36
Price (USD)
Price: 149.00
Online price: 126.65
Your price:
Quantity
-

Chemical Identifiers

CAS
89-65-6
Form
crystals
Appearance (Color)
White to pale yellow
Assay (Titration)
> 98.5%
Optical Rotation
-16.0 to -19.0 deg ( c=10, water)

Description

D-(-)-Isoascorbic acid is used as antioxidant especially in brewing industry, reducing agent in photography. And it is also used in food industry, as food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text