Thermo Scientific Chemicals

Magnesium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation, Thermo Scientific Chemicals

Catalog number: 036427.KU
5 Ea., Each
Thermo Scientific Chemicals

Magnesium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation, Thermo Scientific Chemicals

Catalog number: 036427.KU
5 Ea., Each
Quantity
Catalog number: 036427.KU
also known as 036427-KU
Price (USD)
Price: 440.00
Online price: 373.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1309-48-4
IUPAC Name
oxomagnesium
Molecular Formula
MgO
InChI Key
CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES
O=[Mg]
Width
9 - 11 mm
Height
0.4 -0.6 mm
Form
Substrate
Comment
100 orientation
Comment
Polished one side

Description

A useful laboratory reagent, and a good reference white color for colorimetry. The interaction of magnesium oxide substrates with nickel ferrite films has been investigated using a wide range of electron optical techniques. Considerable interdiffusion is observed, which is likely to alter the magnetic properties of these films. Epitaxial ferrite-spinel layer is formed on magnesium oxide substrate. Additionally, Magnesium oxide in nanoparticles dispersed in alcohol have been reported to inhibit hydrolytic and oxidative degradation of paper documents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text