Thermo Scientific Chemicals

Starch, modified, insolubles 0.01% max, Thermo Scientific Chemicals

Catalog number: 036673.36
500 g, Each
Thermo Scientific Chemicals

Starch, modified, insolubles 0.01% max, Thermo Scientific Chemicals

Catalog number: 036673.36
500 g, Each
Quantity
Catalog number: 036673.36
also known as 036673-36
Price (USD)
Price: 160.00
Online price: 136.65
Your price:
Quantity
-

Chemical Identifiers

CAS
9005-84-9
IUPAC Name
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
GUBGYTABKSRVRQ-ASMJPISFSA-N
SMILES
OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Clarity
Slightly hazy solution
Insoluble material
≤0.01 wt%
Appearance (Color)
White

Description

Starch, modified is used in the production of facial cleanser, oil controller, sunscreen cream, moisturizer, toners/astringents, facial moisturizer/treatment and anti-aging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text