Lithiummetaborat, ACS, 98.0 bis 102.0 %, Thermo Scientific Chemicals
Lithiummetaborat, ACS, 98.0 bis 102.0 %, Thermo Scientific Chemicals
Lithiummetaborat, ACS, 98.0 bis 102.0 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithiummetaborat, ACS, 98.0 bis 102.0 %, Thermo Scientific Chemicals

CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
Katalognummer 036760.18
auch als 036760-18 bezeichnet
Preis (EUR)
101,00
Each
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
101,00
Each
Chemikalien-Kennzeichnungen
CAS13453-69-5
IUPAC Nametetraboron heptahydrate dilithium
Molecular FormulaB4H14Li2O7
InChI KeyDPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)98.0-102.0% BLiO₂
Impurity contentAl 0.001%
Impurity contentFe 0.002%
Impurity contentMg 5ppm
Impurity contentPhosphorus compounds (as PO₄) 0.004%
Mehr anzeigen
It acts as a support for antioxidant property in human and animal tissues. Used in non-linear optic materials and cellular phones. It is used for measuring total elements in soil, geological, plant and other natural organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only