Thermo Scientific Chemicals

Ruthenium, 0.5% on granular carbon, Thermo Scientific Chemicals

Catalog number: 038332.14
25 g, Each
Thermo Scientific Chemicals

Ruthenium, 0.5% on granular carbon, Thermo Scientific Chemicals

Catalog number: 038332.14
25 g, Each
Quantity
Catalog number: 038332.14
also known as 038332-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7440-18-8
IUPAC Name
ruthenium
Molecular Formula
Ru
InChI Key
KJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES
[Ru]
Assay from Supplier's CofA
0.45-0.55%
Particle Size
4-12 mesh
Appearance (Color)
Black
Total Metal Impurities
≤0.01%
Surface Area
900-1100 m²/g

Description

Ruthenium is used in wear-resistant electrical contacts and the production of thick-film resistors. Its use in some platinum alloys, and as a catalyst. It is a most effective hardeners for platinum and palladium. It is also used in some advanced high-temperature single-crystal super alloys, with applications including the turbine blades in jet engines and fountain pen nibs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text