Strontiumsulfat, Reagenzien-Gütegrad, Thermo Scientific Chemicals
Strontiumsulfat, Reagenzien-Gütegrad, Thermo Scientific Chemicals
Strontiumsulfat, Reagenzien-Gütegrad, Thermo Scientific Chemicals
Strontiumsulfat, Reagenzien-Gütegrad, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontiumsulfat, Reagenzien-Gütegrad, Thermo Scientific Chemicals

CAS: 7759-02-6 | O4SSr | 183.68 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
Katalognummer 038373.14
auch als 038373-14 bezeichnet
Preis (EUR)
34,50
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
34,50
Each
Chemikalien-Kennzeichnungen
CAS7759-02-6
IUPAC Namestrontium(2+) sulfate
Molecular FormulaO4SSr
InChI KeyUBXAKNTVXQMEAG-UHFFFAOYSA-L
SMILES[Sr++].[O-]S([O-])(=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Impurity content0.5% max
Strontium sulfate is used as a precursor to other strontium compounds and as ceramic precursor. Its application in nitrate for use in pyrotechnics. Used as a fining agent in crystal glass. It is also used in powder coating, liquid paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only