Dichlormethan, Thermo Scientific Chemicals
Dichlormethan, Thermo Scientific Chemicals
Dichlormethan, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichlormethan, Thermo Scientific Chemicals

Maximaler Gehalt an Verunreinigungen: Farbe (APHA) 10, Restmenge nach Verdampfung 0.002 %, Titrierbare Säuremenge 0.0003 meq/g, freie Halogene P.T., H2 O0.02 %
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 mL
1 L
4 x 1 L
Katalognummer 039116.M1
auch als 039116-M1 bezeichnet
Preis (EUR)
-
Menge:
4 x 1 L
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Free acid (titration)≤0.0003meq/g
Residue on Evaporation≤0.002%
FormLiquid
Assay (GC)>99.5%
Residual water≤0.02%
Mehr anzeigen
Dichloromethane is a chlorinated hydrocarbon that has been employed as a pharamacuetical applications. Mainly, it's utilization as a solvent in manufacturing and food engineering. It is too useful in the drinking bird heat engine. It was the solvent of choice for caffeine extraction. It is likewise employed as a solvent and paint remover. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only