Thermo Scientific Chemicals

2-Butanone, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 039119.K7
4 L, Each
Thermo Scientific Chemicals

2-Butanone, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 039119.K7
4 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 039119.K7
also known as 039119-K7
Price (USD)
Price: 131.00
Your price:
Quantity
-

Chemical Identifiers

CAS
78-93-3
IUPAC Name
butan-2-one
Molecular Formula
C4H8O
InChI Key
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
SMILES
CCC(C)=O
Color (APHA)
≤15
Residue on Evaporation
≤0.0025%
Free acid (titration)
≤0.0005meq/g
Form
Liquid
Water Content (Karl Fischer Titration)
≤0.20%

Description

2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text