Thermo Scientific Chemicals

Potassium metavanadate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039501.14
25 g, Each
Thermo Scientific Chemicals

Potassium metavanadate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039501.14
25 g, Each
Quantity
Catalog number: 039501.14
also known as 039501-14
Price (USD)
Price: 82.40
Online price: 70.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13769-43-2
IUPAC Name
potassium vanadiumoylolate
Molecular Formula
KO3V
InChI Key
BQFYGYJPBUKISI-UHFFFAOYSA-N
SMILES
[K+].[O-][V](=O)=O
Total Metal Impurities
0.1% max.

Description

Employed as corrosion inhibitors and anti-scaling agents. Used widely in chemical fertilizer industries, glass and ceramic industries, pharmaceutical industries. Also used in the process of pigment production for plastics, enamels, ceramics. Used for industrial gas desulphuration and H2S adsorption and elimination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text