Kaliummethansulfonat, 99 %, Thermo Scientific Chemicals
Kaliummethansulfonat, 99 %, Thermo Scientific Chemicals
Kaliummethansulfonat, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kaliummethansulfonat, 99 %, Thermo Scientific Chemicals

CAS: 2386-56-3 | CH3KO3S | 134.19 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
500 g
Katalognummer 039505.14
auch als 039505-14 bezeichnet
Preis (EUR)
67,10
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
67,10
Each
Chemikalien-Kennzeichnungen
CAS2386-56-3
IUPAC Namepotassium methanesulfonate
Molecular FormulaCH3KO3S
InChI KeyXWIJIXWOZCRYEL-UHFFFAOYSA-M
SMILES[K+].CS([O-])(=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White to pale cream
FormPowder or crystalline powder or crystals or lumpy powder
Assay (unspecified)≥98.5%
Used in pharmaceutical, chemical and biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only