Tetrachlorbis-(tetrahydrofuran)-zirkon(IV), Thermo Scientific Chemicals
Tetrachlorbis-(tetrahydrofuran)-zirkon(IV), Thermo Scientific Chemicals
Tetrachlorbis-(tetrahydrofuran)-zirkon(IV), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrachlorbis-(tetrahydrofuran)-zirkon(IV), Thermo Scientific Chemicals

Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
2 g
10 g
Katalognummer 039550.04
auch als 039550-04 bezeichnet
Preis (EUR)
-
Menge:
2 g
Großbestellung oder individuelle Größe anfordern
Specifications
CAS21959-01-3
Chemischer Name oder MaterialTetrachlorobis(tetrahydrofuran)zirconium(IV)
FarbeWeiß
DOT-InformationenGefahrenklasse:8; Verpackungsgruppe:III
Formelmasse377.25
Mehr anzeigen
It is an important organic intermediate and can be used in agrochemicals, paharmaceuticals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only