Titan(IV)-Oxidsulfat-Schwefelsäurehydrat, Thermo Scientific Chemicals
Titan(IV)-Oxidsulfat-Schwefelsäurehydrat, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titan(IV)-Oxidsulfat-Schwefelsäurehydrat, Thermo Scientific Chemicals

CAS: 123334-00-9 | TiOSO4.xH2O + H2SO4
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 g
2 kg
Katalognummer 039591.36
auch als 039591-36 bezeichnet
Preis (EUR)
85,85
Exklusiv online
101,00
Ersparnis 15,15 (15%)
Each
Zum Warenkorb hinzufügen
Menge:
500 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
85,85
Exklusiv online
101,00
Ersparnis 15,15 (15%)
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS123334-00-9
SpecificationsSpecification SheetDatenblatt
Assay from Suppliers CofAFree-Sulfuric Acid: Report Value
Assay from Suppliers CofAWater: Report Value
Assay from Suppliers CofATitanium Oxysulfate: Report value
Titanium(IV) oxide sulfate sulfuric acid hydrate is widely used as a pigment in paints and dyes. It is also useful in the preparation of stable amorphous titanium oxide hollow microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only