Zinn(II)-trifluormethansulfonat, 97 %, Thermo Scientific Chemicals
Zinn(II)-trifluormethansulfonat, 97 %, Thermo Scientific Chemicals
Zinn(II)-trifluormethansulfonat, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinn(II)-trifluormethansulfonat, 97 %, Thermo Scientific Chemicals

CAS: 62086-04-8 | CH4F3O3SSn+ | 271.805 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
Katalognummer 039603.03
auch als 039603-03 bezeichnet
Preis (EUR)
50,90
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
50,90
Each
Specifications
CAS62086-04-8
Chemischer Name oder MaterialTin(II) trifluoromethanesulfonate
DOT-InformationenGefahrenklasse:8; Verpackungsgruppe:III
Formelmasse416.82
Gesundheitsgefahr 1Gehäuse H314-H500
Mehr anzeigen
Tin(II) trifluoromethanesulfonate is a mild Lewis acid used for stereoselective aldol and Michael reactions; [2,3]-Wittig and Ireland-Claisen rearrangements, Horner-Wadsworth-Emmons reactions, asymmetric [3 + 2] cycloaddition, ring opening reactions and Friedel-Crafts reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only