Copper slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.996% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.996% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 039675.22
also known as 039675-22
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
Copper slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.996% (metals basis), Thermo Scientific Chemicals
Catalog number039675.22
Price (USD)/ Each
114.00
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSlug, 6.35 mm dia., 6.35 mm length
Appearance (Color)Copper
Total Metal Impurities≤0.004%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only