Ethylendiamin-Tetraessigsäure-Dinatriumsalz, 0.100N (0.050M) Standardlösung, Thermo Scientific Chemicals
Ethylendiamin-Tetraessigsäure-Dinatriumsalz, 0.100N (0.050M) Standardlösung, Thermo Scientific Chemicals
Ethylendiamin-Tetraessigsäure-Dinatriumsalz, 0.100N (0.050M) Standardlösung, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylendiamin-Tetraessigsäure-Dinatriumsalz, 0.100N (0.050M) Standardlösung, Thermo Scientific Chemicals

CAS: 139-33-3 | C10H14N2Na2O8 | 336.21 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 L
4 L
Katalognummer 039781.K7
auch als 039781-K7 bezeichnet
Preis (EUR)
91,50
Each
Menge:
4 L
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
91,50
Each
Chemikalien-Kennzeichnungen
CAS139-33-3
IUPAC Namedisodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Molecular FormulaC10H14N2Na2O8
InChI KeyZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Clear colorless
FormLiquid
Concentration0.100N ± 0.19%
It is widely used in textile industry. Usually applied to dissolve limescale. It is applied in textile industry, pulp and paper industry and also in chelation therapy. In cosmetics it acts as a sequestering agent. It acts as a corrosion inhibitor to carbon steel in the industries. It also acts as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only