Triethylphosphat, 99+%, Thermo Scientific Chemicals
Triethylphosphat, 99+%, Thermo Scientific Chemicals
Triethylphosphat, 99+%, Thermo Scientific Chemicals
Triethylphosphat, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylphosphat, 99+%, Thermo Scientific Chemicals

CAS: 78-40-0 | C6H15O4P | 182.16 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
500 g
Katalognummer 040001.14
auch als 040001-14 bezeichnet
Preis (EUR)
34,70
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
34,70
Each
Specifications
CAS78-40-0
ChEBICHEBI:45927
Chemischer Name oder MaterialTriethyl phosphate
Dichte1.072 g/mL
EINECS-Nummer201-114-5
Mehr anzeigen
Triethyl phosphate is used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only